Learn More
HiCK-1 Cytokine Positive Control Cells, BD
Brand:  BD Biosciences 555061
Product Code. 15827988
  • ONLINE EXCLUSIVE PRICE
    £302.00
    £242.93 / Each
    Save £59.07
    19% Off
Description
ã5 x 106 activated human PBMCs (20 test samples at 2.5 x 105 cells/test) fixed and suspended in 1mL (10% dimethylsulfoxide and 90% fetal bovine serum)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5x106 cells/ml