missing translation for 'onlineSavingsMsg'
Learn More

HIC5/TGFB1I1 Antibody, Novus Biologicals™

Product Code. 18384378 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384378 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384378 Supplier Novus Biologicals Supplier No. H00007041D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HIC5/TGFB1I1 Polyclonal antibody specifically detects HIC5/TGFB1I1 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIC5/TGFB1I1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_057011.2
Gene Alias Androgen receptor coactivator 55 kDa protein, androgen receptor coactivator ARA55, Androgen receptor-associated protein of 55 kDa, ARA55Hydrogen peroxide-inducible clone 5 protein, HIC5, HIC-5, transforming growth factor beta 1 induced transcript 1, transforming growth factor beta-1-induced transcript 1 protein, TSC-5
Host Species Rabbit
Immunogen TGFB1I1 (NP_057011.2, 1 a.a. - 444 a.a.) full-length human protein. MPRSGAPKERPAEPLTPPPSYGHQPQTGSGESSGASGDKDHLYSTVCKPRSPKPAAPAAPPFSSSSGVLGTGLCELDRLLQELNATQFNITDEIMSQFPSSKVASGEQKEDQSEDKKRPSLPSSPSPGLPKASATSATLELDRLMASLSDFRVQNHLPASGPTQPPVVSSTNEGSPSPPEPTGKGSLDTMLGLLQSDLSRRGVPTQAKGLCGSCNKPIAGQVVTALGRAWHPEHFVCGGCSTALGGSSFFEKDGAPFCPECYFERFSPRCGFCNQPIRHKMVTALGTHWHPEHFCCVSCGEPFGDEGFHEREGRPYCRRDFLQLFAPRCQGCQGPILDNYISALSALWHPDCFVCRECFAPFSGGSFFEHEGRPLCENHFHARRGSLCATCGLPVTGRCVSALGRRFHPDHFTCTFCLRPLTKGSFQERAGKPYCQPCFLKLFG
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 7041
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.