missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528839
This item is not returnable.
View return policy
Description
Recommended positive controls: Hic-5 tansfected 293T cell. Predicted reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a coactivator of the androgen receptor, a transcription factor which is activated by androgen and has a key role in male sexual differentiation. The encoded protein is thought to regulate androgen receptor activity and may have a role to play in the treatment of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| HIC5 | |
| Polyclonal | |
| Unconjugated | |
| Tgfb1i1 | |
| Androgen receptor coactivator 55 kDa protein; androgen receptor coactivator ARA55; androgen receptor-associated protein of 55 kDa; Ara55; Hic5; hic-5; hydrogen peroxide-inducible clone 5 protein; hydrogen peroxide-inducible clone-5; TGF beta-stimulated clone 5; Tgfb1i1; transforming growth factor beta 1 induced transcript 1; transforming growth factor beta-1-induced transcript 1 protein; TSC-5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7041 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.79 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| O43294 | |
| Tgfb1i1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human HIC5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction