missing translation for 'onlineSavingsMsg'
Learn More

HIC2 Antibody (PCRP-HIC2-2F8), DyLight 350, Novus Biologicals™

Product Code. 18374516 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374516 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374516 Supplier Novus Biologicals Supplier No. NBP314235UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HIC2 Monoclonal antibody specifically detects HIC2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIC2
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-HIC2-2F8
Conjugate DyLight 350
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias HIC1-related gene on chromosome 22 protein, Hic-2, Hic-3, HRG22KIAA1020hic-2, hypermethylated in cancer 2, hypermethylated in cancer 2 protein, ZBTB30hic-3, Zinc finger and BTB domain-containing protein 30, ZNF907
Host Species Mouse
Immunogen Recombinant full-length human HIC2protein (Uniprot: Q96JB3)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23119
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.