missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIC2 Antibody (PCRP-HIC2-2F8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14235B
This item is not returnable.
View return policy
Description
HIC2 Monoclonal antibody specifically detects HIC2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| HIC2 | |
| Monoclonal | |
| Biotin | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 23119 | |
| Store at 4°C in the dark. | |
| Purified |
| Flow Cytometry, ELISA, Immunofluorescence | |
| PCRP-HIC2-2F8 | |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| HIC1-related gene on chromosome 22 protein, Hic-2, Hic-3, HRG22KIAA1020hic-2, hypermethylated in cancer 2, hypermethylated in cancer 2 protein, ZBTB30hic-3, Zinc finger and BTB domain-containing protein 30, ZNF907 | |
| Recombinant full-length human HIC2protein (Uniprot: Q96JB3) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction