missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIC1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24949-1-AP-150UL
This item is not returnable.
View return policy
Specifications
| HIC1 | |
| Polyclonal | |
| Unconjugated | |
| HIC1 | |
| hic 1, HIC1, hypermethylated in cancer 1, ZBTB29 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15248, 3090 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14526, Q9R1Y5 | |
| Hic1 | |
| HIC1 Fusion Protein Ag21849 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction