missing translation for 'onlineSavingsMsg'
Learn More

FTM Technologies™ HI1P Portable Fume Hood

Product Code. 11798054
Change view
Click to view available options
Flow Rate:
50 to 430 m3/h
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Flow Rate Unit Size
11798054 50 to 430 m3/h Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11798054 Supplier FTM Technologies™ Supplier No. HI1P
  • ONLINE EXCLUSIVE PRICE
    £1635.00
    £1147.03 / Pack of 1
    Save £487.97
    29% Off
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Protects the face against the risk of inhalation and splashes of hazardous materials. FTM Technologies™ HI1P Portable Fume Hoods have a height-adjustable illuminated visor.

  • Easy to move
  • Translucent angled visor
  • Polypropylene housing, brushless motor
  • Use as a recirculating (ductless) fume hood or with an external output/pre-filters

Specifications

Dimensions (L x W x H) 350 x 500 x 470 mm
Flow Rate 50 to 430 m3/h
Voltage 230 V
Weight (Metric) 6 kg
Frequency 50 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.