missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Hi-Di™ Formamide

Product Code. 16325322 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16325322 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 16325322 Supplier Applied Biosystems™ Supplier No. 4401457

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

The smaller configuration(5mL) minimizes the number of freeze/thaw cycles compared to the larger 25mL bottle.

Highly deionized (Hi-Di) formamide is used to resuspend samples before electrokinetic injection in capillary electrophoresis systems.

Order Info

Each

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mL
Physical Form Liquid
Product Line Hi-Di
Molecular Formula CH3NO
MDL Number MFCD00007941
InChI Key ZHNUHDYFZUAESO-UHFFFAOYSA-N
SMILES NC=O
IUPAC Name formamide
Molecular Weight (g/mol) 45.04

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.