missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HHV8-ORF74 Antibody (462510), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5264C
This item is not returnable.
View return policy
Description
HHV8-ORF74 Monoclonal antibody specifically detects HHV8-ORF74 in Virus samples. It is validated for Flow Cytometry, CyTOF
Specifications
| HHV8-ORF74 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| NS0 mouse myeloma cell line transfected with viral HHV8-ORF74, Met1-Thr342, Accession # AAB51506 | |
| 0.1 mL | |
| Primary | |
| Virus | |
| Purified |
| Flow Cytometry, CyTOF | |
| 462510 | |
| Flow Cytometry, CyTOF-ready | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4961460 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction