missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ HHV-8 ORF K4 Recombinant Protein, PeproTech®

Product Code. 17892203
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17892203

Brand: Gibco™ 35003100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Viral MIP-2 is a 7.9 kDa protein consisting of 70 amino acids, including the four highly conserved cysteine residues present in CC chemokines. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Viral MIP-2 (vMIP-2) is a chemokine analog encoded by the human herpes virus, and has been shown to have antagonist activity towards several chemokine receptors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q98157
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 4961514
Molecular Weight (g/mol) 7.9 kDa
Name HHV-8 ORF K4
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Common Name HHV-8 ORF K4
Gene Symbol K4
Biological Activity Determined by the inhibitory effect on monocyte migration response to human MIP-1 alpha using a concentration range of 1.0 ug-10.0 ug/ml of viral MIP-2 will inhibit 25 ng/ml of human MIP-1 alpha.
Conjugate Unconjugated
Recombinant Recombinant
Sequence LGASWHRPDK CCLGYQKRPL PQVLLSSWYP TSQLCSKPGV IFLTKRGRQV CADKSKDWVK KLMQQLPVTA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.