Learn More
Invitrogen™ HHV-6 90kDa Glycoprotein Monoclonal Antibody (F25I)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17045
Description
MA1-7045 detects the Human Herpes virus, type 6. This antibody is specific for a 90 kDa glycoprotein of HHV6. MA1-7045 has been successfully used in immunoprecipitation, immunohistochemistry and Western blot procedures. The MA1-7045 immunogen is Human Herpes Virus type 6 lysate.
Specifications
| HHV-6 90kDa Glycoprotein | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2 |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| F25I | |
| Unconjugated | |
| HHV-6; HHV-6 gp | |
| Human Herpes Virus type 6 lysate. | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.