missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HHV-1 US3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567986
This item is not returnable.
View return policy
Description
HHV-1 US3 Polyclonal Antibody for Western Blot
US3 is a multifunctional serine/threonine kinase that plays a role in several processes including egress of virus particles from the nucleus, modulation of the actin cytoskeleton and inhibition of apoptosis. It phosphorylates UL31 and UL34, two critical regulators of capsid budding from nucleus to endoplasmic reticulum, thereby facilitating virion egress. US3 modulates and redistributes host components of the nuclear envelope, including LMNA, emerin/EMD and the nuclear matrix protein MATR3. It also phosphorylates envelope glycoprotein B (gB), probably to direct it to the cell surface, and promotes virus intracellular spread by restructuring host cell cytoskeleton. US3 blocks host apoptosis to extend cell survival and allow efficient viral replication. Promotes viral gene expression by phosphorylating host HDAC2 to reduce viral genome silencing.
Specifications
| HHV-1 US3 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.4 | |
| Synthesized peptide derived from human KR1_HHV11. | |
| 100 μL | |
| Primary | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction