missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hHR23b Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-89699
This item is not returnable.
View return policy
Description
hHR23b Polyclonal specifically detects hHR23b in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| hHR23b | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| RAD23B | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VIEALRASFNNPDRAVEYLLMGIPGDRESQAVVDPPQAASTGAPQSSAVAAAAATTTATTTTTSSGGHPLEFLRNQPQFQQMRQIIQQNP | |
| 0.1 mL | |
| Apoptosis, Cancer, Core ESC Like Genes, DNA Repair, Nucleotide Excision Repair, Stem Cell Markers, Tumor Suppressors | |
| 5887 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.1mg/mL | |
| Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| HHR23B, HR23BUV excision repair protein RAD23 homolog B, p58, RAD23 (S. cerevisiae) homolog B, RAD23 homolog B (S. cerevisiae), RAD23, yeast homolog of, B, XP-C repair complementing complex 58 kDa, XP-C repair complementing protein, XP-C repair-complementing complex 58 kDa protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction