missing translation for 'onlineSavingsMsg'
Learn More

hHpr1-p84-Thoc1 Rabbit anti-Human, Mouse, Rat, Clone: 6E3A7, Novus Biologicals™

Product Code. 18389212
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18389212 20 μg 20µL
18307004 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18389212 Supplier Bio-Techne Supplier No. NBP31680020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

hHpr1-p84-Thoc1 Monoclonal antibody specifically detects hHpr1-p84-Thoc1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen hHpr1-p84-Thoc1
Applications Western Blot
Classification Monoclonal
Clone 6E3A7
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias HPR1THO complex subunit 1, hTREX84, Nuclear matrix protein p84, P84, P84N5, THO complex 1, tho1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 350-450 of human hHpr1-p84-Thoc1 (Q96FV9).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, DNA Repair, DNA replication Transcription Translation and Splicing, Nuclear Envelope Markers
Primary or Secondary Primary
Gene ID (Entrez) 9984
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.