missing translation for 'onlineSavingsMsg'
Learn More

HHEX Antibody (OTI3E6), DyLight 594, Novus Biologicals™

Product Code. 18432593 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432593 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432593 Supplier Novus Biologicals Supplier No. NBP270890DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HHEX Monoclonal antibody specifically detects HHEX in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HHEX
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone OTI3E6
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias hematopoietically expressed homeobox, HEXHMPH, Homeobox protein HEX, Homeobox protein PRH, HOX11L-PEN, PRHhematopoietically-expressed homeobox protein HHEX, PRHXhomeobox, hematopoietically expressed, proline-rich homeodomain-containing transcription factor
Host Species Mouse
Immunogen Recombinant fragment expressed in E.coli corresponding to amino acids 100-270 of human HHEX.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 3087
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.