missing translation for 'onlineSavingsMsg'
Learn More

HHEX Antibody (2C8), Novus Biologicals™

Product Code. 18338037 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338037 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338037 Supplier Novus Biologicals Supplier No. H00003087M12

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HHEX Monoclonal antibody specifically detects HHEX in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen HHEX
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2C8
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_002720
Gene Alias hematopoietically expressed homeobox, HEXHMPH, Homeobox protein HEX, Homeobox protein PRH, HOX11L-PEN, PRHhematopoietically-expressed homeobox protein HHEX, PRHXhomeobox, hematopoietically expressed, proline-rich homeodomain-containing transcription factor
Host Species Mouse
Immunogen HHEX (NP_002720, 133 a.a. ~ 237 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PLHKRKGGQVRFSNDQTIELEKKFETQKYLSPPERKRLAKMLQLSERQVKTWFQNRRAKWRRLKQENPQSNKKEELESLDSSCDQRQDLPSEQNKGASLDSSQCS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Developmental Biology
Primary or Secondary Primary
Gene ID (Entrez) 3087
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.