missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HHATL Polyclonal Antibody

HHATL Polyclonal Antibody
Brand: Invitrogen PA534355
This item is not returnable.
View return policy
Description
HHATL Polyclonal Antibody for Western Blot, IHC
Specifications
| HHATL | |
| Polyclonal | |
| Unconjugated | |
| HHATL | |
| 1110011D13Rik; C3orf3; Glycerol uptake/transporter homolog; GUP1; GUP1 glycerol uptake/transporter homolog; Gup1, glycerol uptake/transporter homolog; hedgehog acyltransferase-like; hedgehog acyltransferase-like protein; HHATL; KIAA1173; MBOAT3; membrane bound O-acyltransferase domain containing 3; MSTP002; OACT3; Protein-cysteine N-palmitoyltransferase HHAT-like protein; RGD1311911 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 301073, 57467, 74770 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9D1G3, Q9HCP6 | |
| HHATL | |
| A 19 amino acid synthetic peptide near the amino terminus of human HHATL. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction