missing translation for 'onlineSavingsMsg'
Learn More

HHATL Antibody, mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18412806 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412806 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412806 Supplier Novus Biologicals Supplier No. NBP281952MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HHATL Polyclonal antibody specifically detects HHATL in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HHATL
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C3orf3, chromosome 3 open reading frame 3, glycerol uptake/transporter homolog, GUP1, hedgehog acyltransferase-like, Hedgehog acyltransferase-like protein, KIAA1173GUP1, glycerol uptake/transporter homolog (yeast), MBOAT3, membrane bound O-acyltransferase domain containing 3, protein-cysteine N-palmitoyltransferase HHAT-like protein
Host Species Rabbit
Immunogen HHATL antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human HHATL. The immunogen is located within the first 50 amino acids of HHATL. Amino Acid Squence: EASQDGAHRKAFRESVRPG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 57467
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.