missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HGSNAT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556197
This item is not returnable.
View return policy
Description
Immunogen sequence: QALLLIHNEL LWTNLTVYWK SECCYHCLFQ VLVNVPQSPK AGKPSAAAAS VSTQHGSILQ LNDTLEEKEV CRLEYRFGEF GNYSLLVKNI HNGVSEIACD LAVN Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 23%.
This gene encodes a lysosomal acetyltransferase, which is one of several enzymes involved in the lysosomal degradation of heparin sulfate. Mutations in this gene are associated with Sanfilippo syndrome C, one type of the lysosomal storage disease mucopolysaccaridosis III, which results from impaired degradation of heparan sulfate.
Specifications
| HGSNAT | |
| Polyclonal | |
| Unconjugated | |
| HGSNAT | |
| Heparan-alpha-glucosaminide N-acetyltransferase; HGNAT; HGSNAT; MPS3C; RP73; TMEM76; transmembrane protein 76 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 138050 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q68CP4 | |
| HGSNAT | |
| Recombinant protein corresponding to Human HGSNAT. Recombinant protein control fragment (Product #RP-90544). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction