missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HGS Recombinant Rabbit Monoclonal Antibody (HL1730)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547159
This item is not returnable.
View return policy
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
HGS is involved in intracellular signal transduction mediated by cytokines and growth factors. When associated with STAM, it suppresses DNA signaling upon stimulation by IL-2 and GM-CSF. It could be a direct effector of PI3-kinase in vesicular pathway via early endosomes and may regulate trafficking to early and late endosomes by recruiting clathrin. HGS may concentrate ubiquitinated receptors within clathrin-coated regions. It is involved in down-regulation of receptor tyrosine kinase via multivesicular body (MVBs) when complexed with STAM. This complex binds ubiquitin and acts as sorting machinery that recognizes ubiquitinated receptors and transfers them to further sequential lysosomal sorting/trafficking processes. HGS may contribute to the efficient recruitment of SMADs to the activin receptor complex.
Specifications
| HGS | |
| Recombinant Monoclonal | |
| 2 mg/mL | |
| PBS with no preservative | |
| O14964 | |
| HGS | |
| Recombinant protein encompassing a sequence within the C-terminus region of human HGS. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Canine, Feline | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| HL1730 | |
| Unconjugated | |
| HGS | |
| hepatocyte growth factor-regulated tyrosine kinase substrate; HGF-regulated tyrosine kinase substrate; HGNC:4897; Hgr; Hgs; Hrs; Hrs2; human growth factor-regulated tyrosine kinase substrate; protein pp110; SNAP-25-interacting protein Hrs-2; ZFYVE8 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 101088702, 483367, 9146 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction