missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGS Antibody (OTI3H7), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71418AF594
This item is not returnable.
View return policy
Description
HGS Monoclonal specifically detects HGS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| HGS | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HGS | |
| Full length recombinant protein of human HGS (NP_004703) produced in HEK293T cells. | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI3H7 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| FLJ39878, hepatocyte growth factor-regulated tyrosine kinase substrate, Hrs, HRSFLJ11421, human growth factor-regulated tyrosine kinase substrate, Protein pp110, Vps27, ZFYVE8 | |
| Mouse | |
| 0.1 mL | |
| 9146 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction