missing translation for 'onlineSavingsMsg'
Learn More

HGK/MAP4K4 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18738450 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18738450 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18738450 Supplier Novus Biologicals Supplier No. NB10074630AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HGK/MAP4K4 Polyclonal antibody specifically detects HGK/MAP4K4 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen HGK/MAP4K4
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 2.7.11, EC 2.7.11.1, FLH21957, FLJ10410, hepatocyte progenitor kinase-like/germinal center kinase-like kinase, HGKFLJ20373, HPK/GCK-like kinase HGK, KIAA0687, MAPK/ERK kinase kinase kinase 4, MEK kinase kinase 4, MEKKK 4, mitogen-activated protein kinase kinase kinase kinase 4, Nck-interacting kinase, NIKFLJ90111
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 850 and 900 of human MAPK/ERK kinase kinase kinase 4 (hepatocyte progenitor kinase-like/germinal center kinase-like kinase) using the numbering given in entry AAO32626.1 (GeneID 94
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 9448
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.