missing translation for 'onlineSavingsMsg'
Learn More

HGK/MAP4K4 Antibody (3D9), Novus Biologicals™

Product Code. 18339828 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339828 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339828 Supplier Novus Biologicals Supplier No. H00009448M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HGK/MAP4K4 Monoclonal antibody specifically detects HGK/MAP4K4 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen HGK/MAP4K4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 3D9
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_663719
Gene Alias EC 2.7.11, EC 2.7.11.1, FLH21957, FLJ10410, hepatocyte progenitor kinase-like/germinal center kinase-like kinase, HGKFLJ20373, HPK/GCK-like kinase HGK, KIAA0687, MAPK/ERK kinase kinase kinase 4, MEK kinase kinase 4, MEKKK 4, mitogen-activated protein kinase kinase kinase kinase 4, Nck-interacting kinase, NIKFLJ90111
Host Species Mouse
Immunogen MAP4K4 (NP_663719, 611 a.a. ~ 710 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VHPALQRPAEPQVQWSHLASLKNNVSPVSRSHSFSDPSPKFAHHHLRSQDPCPPSRSEVLSQSSDSKSEAPDPTQKAWSRSDSDEVPPRVPVRTTSRSPV
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 9448
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.