missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGH1 Polyclonal Antibody
HGH1 Polyclonal Antibody
Brand: Invitrogen PA571387
This item is not returnable.
View return policy
Description
HGH1 Polyclonal Antibody for Western Blot
Specifications
| HGH1 | |
| Polyclonal | |
| Unconjugated | |
| HGH1 | |
| brain protein 16; brain protein 16-like; Brp16; BRP16L; C8orf30A; C8orf30B; D15Ertd741e; Fam203a; FAM203B; family with sequence similarity 203, member A; family with sequence similarity 203, member B; Hgh1; HGH1 homolog; MNCb-5873; protein FAM203A; protein FAM203B; protein HGH1 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 59053 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8C3I8 | |
| HGH1 | |
| synthetic peptide directed towards the middle region of mouse HGH1. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction