missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HGH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549992
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total HGH1 protein.
Hgh1 is a protein coding gene.
Specifications
| HGH1 | |
| Polyclonal | |
| Unconjugated | |
| HGH1 | |
| brain protein 16; brain protein 16-like; Brp16; BRP16L; C8orf30A; C8orf30B; D15Ertd741e; Fam203a; FAM203B; family with sequence similarity 203, member A; family with sequence similarity 203, member B; Hgh1; HGH1 homolog; MNCb-5873; protein FAM203A; protein FAM203B; protein HGH1 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51236 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BTY7 | |
| HGH1 | |
| Synthesized peptide derived from internal of human BRP16. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction