missing translation for 'onlineSavingsMsg'
Learn More

HGH Fragment 176-191 Trifluoroacetic acid (>90%), TRC

Product Code. 30006359
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006359 1 mg 1mg
30134341 5 mg 5mg
30135120 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006359 Supplier Toronto Research Chemicals Supplier No. TRCH9997731MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66004-57-7
Synonym 1,2-Dithia-5,8,11,14,17,20,23-heptaazacyclohexacosane Cyclic Peptide Derivative Trifluoroacetic acid∣HGH FRAG 176-191 Trifluoroacetic acid∣L-phenylalanyl-L-leucyl-L-arginyl-L-isoleucyl-L-valyl-L-glutaminyl-L-cysteinyl-L-arginyl-L-seryl-L-valyl-L-α-glutamylglycyl-L-seryl-L-cysteinylglycyl-L-phenylalanine-Cyclic (7→14)-Disulfide Trifluoroacetic acid;

Specifications

Format Neat
Molecular Formula C78H123N23O22S2
Molecular Weight (g/mol) 1799.0811402, 1799.10
InChI Key LSTCHEVTSMJOCE-YWWHUHEESA-N
SMILES CC[C@H](C)[C@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](N)CC1=CC=CC=C1)C(=O)N[C@@H](C(C)C)C(=O)OC(=O)CC[C@H](NC(=O)[C@@H](NC(=O)[C@H](CO)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](NC(=O)[C@@H](N)CCC(N)=O)C=S)C(C)C)C(=O)NCC(=O)OC(=O)[C@H](CC1=CC=CC=C1)NC(=O)CNC(=O)[C@H](CS)NC(=O)[C@@H](N)CO
IUPAC Name (2S)-2-[(2S,3S)-2-[(2S)-2-[(2S)-2-[(2S)-2-amino-3-phenylpropanamido]-4-methylpentanamido]-5-carbamimidamidopentanamido]-3-methylpentanamido]-3-methylbutanoyl (4S)-4-[(2-{[(2S)-2-{2-[(2R)-2-[(2S)-2-amino-3-hydroxypropanamido]-3-sulfanylpropanamido]acetamido}-3-phenylpropanoyl]oxy}-2-oxoethyl)carbamoyl]-4-[(2S)-2-[(2S)-2-[(2S)-2-[(2R)-2-[(2S)-2-amino-4-carbamoylbutanamido]-2-(methanethioyl)acetamido]-5-carbamimidamidopentanamido]-3-hydroxypropanamido]-3-methylbutanamido]butanoate
Quantity 1 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.