missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HGF61 Glass Fiber Tape

Product Code. 15979170
Click to view available options
Unit Size:
1 pack
This item is not returnable. View return policy

Product Code. 15979170

Brand: Cytiva 18306236

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provides strength, efficiency, reliability and consistency of data quality in continuous air monitoring.

  • HGF61 glass fiber tape with organic binder has excellent mechanical strength without any fiber shedding.
  • Designed for continuous air particulate monitoring
  • Compatible with most continuous air monitors, and performs well in both dry and humid environments.
  • Each lot is quality tested to the high standard of 99.99% air retention efficiency at 0.3 um.
  • HGF61 glass fiber tape meets global regulatory requirements for continuous air particulate sampling.
  • Suitable for PM1, PM2.5, PM10 and TSP sampling and for beta-attenuation method
  • Provides excellent consitency of data quality in air sampling
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Air Monitor
Material Glass Microfiber
Grade HGF61
Includes Organic and inorganic binder
Quantity 1 Roll
Thickness (Metric) 285 μm
compliance-icons compliance-icons
Product Identifier
  • HGF61, 30 mm x 20 m, 1/PK
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : glass oxide
  • UNKNOWN ACUTE-100 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 100% of components with unknown hazards to the aquatic environment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.