missing translation for 'onlineSavingsMsg'
Learn More

HGF Antibody (MM0326-5D20), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18421496 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421496 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421496 Supplier Novus Biologicals Supplier No. NBP212355MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HGF Monoclonal antibody specifically detects HGF in Human, Bovine samples. It is validated for Western Blot, ELISA, Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen HGF
Applications Western Blot, ELISA, Neutralization
Classification Monoclonal
Clone MM0326-5D20
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, HGFB, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SFhepatopoeitin-A
Host Species Mouse
Immunogen Human recombinant HGF
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 3082
Target Species Human, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.