Learn More
Invitrogen™ HGF Activator Recombinant Rabbit Monoclonal Antibody (101)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA529323
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human HGFA/HGF Activator.
This gene encodes a member of the peptidase S1 protein family. The encoded protein is first synthesized as an inactive single-chain precursor before being activated to a heterodimeric form by endoproteolytic processing. It acts as serine protease that converts hepatocyte growth factor to the active form. Alternative splicing results in multiple transcript variants.
Specifications
| HGF Activator | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q04756 | |
| Hgfac | |
| Recombinant Human HGFA protein (Met1-Ser655). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 101 | |
| Unconjugated | |
| Hgfac | |
| hepatocyte growth factor activator; Hepatocyte growth factor activator long chain; Hepatocyte growth factor activator short chain; HGF activator; HGFA; Hgfac | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3083 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.