missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGF Activator Antibody (101), Janelia Fluor™ 646 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89418JF646
This item is not returnable.
View return policy
Description
HGF Activator Monoclonal antibody specifically detects HGF Activator in Human samples. It is validated for ELISA
Specifications
| HGF Activator | |
| Monoclonal | |
| Janelia Fluor 646 | |
| HGF activator, HGFA, HGFAC | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human HGF Activator (NP_001519.1; Met 1-Ser 655). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 101 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3083 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction