missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HGF Activator Monoclonal antibody specifically detects HGF Activator in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | HGF Activator |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 101 |
| Conjugate | DyLight 680 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | HGF activator, HGFA, HGFAC |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human HGF Activator (NP_001519.1; Met 1-Ser 655). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?