missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16465-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes the enzyme homogentisate 1,2 dioxygenase. This enzyme is involved in the catabolism of the amino acids tyrosine and phenylalanine. Mutations in this gene are the cause of the autosomal recessive metabolism disorder alkaptonuria.Specifications
| HGD | |
| Polyclonal | |
| Unconjugated | |
| HGD | |
| AKU, HGD, HGO, Homogentisate 1,2 dioxygenase, Homogentisate oxygenase, Homogentisic acid oxidase, Homogentisicase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15233, 3081, 360719 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O09173, Q93099 | |
| Hgd | |
| HGD Fusion Protein Ag9544 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction