Learn More
Invitrogen™ HGD Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579360
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human A549 whole cell, rat liver tissue, rat kidney tissue, mouse liver tissue, mouse kidney tissue. IHC: human liver cancer tissue, human prostatic cancer tissue. ICC/IF: CACO-2 cell. Flow: HEPG2 cell.
This gene encodes the enzyme homogentisate 1,2 dioxygenase. This enzyme is involved in the catabolism of the amino acids tyrosine and phenylalanine. Mutations in this gene are the cause of the autosomal recessive metabolism disorder alkaptonuria.
Specifications
| HGD | |
| Polyclonal | |
| Unconjugated | |
| HGD | |
| aku; alkaptonuria; HGD; HGO; homogentisate 1, 2-dioxygenase; homogentisate 1,2-dioxygenase; homogentisate oxidase; homogentisate oxygenase; Homogentisic acid oxidase; homogentisicase | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15233, 3081, 360719 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| O09173, Q93099 | |
| HGD | |
| E. coli-derived human HGD recombinant protein (Position: D374-N445). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.