missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGD Antibody (20D12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26234-50ul
This item is not returnable.
View return policy
Description
HGD Monoclonal antibody specifically detects HGD in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| HGD | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 20D12 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| AKU, EC 1.13.11.5, HGOFLJ94126, homogentisate 1,2-dioxygenase, homogentisate 1,2-dioxygenase (homogentisate oxidase), homogentisate oxidase, Homogentisate oxygenase, Homogentisic acid oxidase, homogentisicase | |
| A synthesized peptide derived from Human HGD [UniProt Q93099] | |
| 50 μL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 3081 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction