missing translation for 'onlineSavingsMsg'
Learn More

HFI-142, MedChemExpress

Product Code. 16497087
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497087 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16497087 Supplier Medchem Express Supplier No. HY1102595mg
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress HFI-142 is an insulin-regulated aminopeptidase (IRAP) inhibitor with a Ki of 2.01 μM.

HFI-142 is an insulin-regulated aminopeptidase (IRAP) inhibitor with a Ki of 2.01 μM.

Specifications

Chemical Name or Material HFI-142
CAS 332164-34-8
Color White
Purity Grade Notes Research
Molecular Formula C17H16N2O4
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (400.23 mM; Need ultrasonic)
SMILES CCOC(C1=C(N)OC2=CC(O)=CC=C2C1C3=CN=CC=C3)=O
Molecular Weight (g/mol) 312.32
Formula Weight 312.32
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.