missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HFE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11758-1-AP-150UL
This item is not returnable.
View return policy
Description
HFE2 is involved in iron metabolism. It may be a component of the signaling pathway which activates hepcidin or it may act as a modulator of hepcidin expression. It could also represent the cellular receptor for hepcidin. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. Defects in this gene are the cause of hemochromatosis type 2A, also called juvenile hemochromatosis (JH). JH is an early-onset autosomal recessive disorder due to severe iron overload resulting in hypogonadotrophic hypogonadism, hepatic fibrosis or cirrhosis and cardiomyopathy, occurring typically before age of 30.Specifications
| HFE2 | |
| Polyclonal | |
| Unconjugated | |
| HFE2 | |
| Hemochromatosis type 2 protein, Hemojuvelin, HFE2, HFE2A, HJV, JH, RGM domain family member C, RGMC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 148738, 310681, 69585 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6ZVN8, Q7TQ32, Q8N7M5 | |
| Hfe2 | |
| HFE2 Fusion Protein Ag2354 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction