Learn More
Invitrogen™ HFE Recombinant Rabbit Monoclonal Antibody (2O7D2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542637
Description
Positive test controls include: HeLa, HT-29, A-549. The target is usually found in the following locations: Cell membrane, Single-pass type I membrane protein. Immunogen sequence: KEFEPKDVLP NGDGTYQGWI TLAVPPGEEQ RYTCQVEHPG LDQPLIVIWE PSPSGTLVIG VISGIAVFVV ILFIGILFII LRKRQGSRGA MGHYVLAERE.
The protein encoded by this gene is a membrane protein that is similar to MHC class I-type proteins and associates with beta2-microglobulin. It is thought that this protein functions to regulate iron absorption by regulating the interaction of the transferrin receptor with transferrin. The iron storage disorder, hereditary haemochromatosis, is a recessive genetic disorder that results from defects in this gene. At least nine alternatively spliced variants have been described for this gene. Additional variants have been found but their full-length nature has not been determined.
Specifications
| HFE | |
| Recombinant Monoclonal | |
| 0.6 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q30201 | |
| HFE | |
| A synthetic peptide corresponding to a sequence within amino acids 249-348 of human HFE (Q30201). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 2O7D2 | |
| Unconjugated | |
| HFE | |
| hemochromatosis; hereditary hemochromatosis protein; hereditary hemochromatosis protein HLA-H; hereditary hemochromatosis protein homolog; Hfe; HFE 1; HFE1; HH; high Fe; HLA H; HLAH; HLA-H; MGC:150812; MGC10379; MGC103790; MHC class I-like protein HFE; Mr2; MVCD7; RT1-CAFE; TFQTL2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3077 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.