missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HFE Antibody (SR1669), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22456-50ul
This item is not returnable.
View return policy
Description
HFE Monoclonal antibody specifically detects HFE in Human samples. It is validated for Western Blot
Specifications
| HFE | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1669 | |
| Western Blot 1:500-1:2000 | |
| dJ221C16.10.1, hemochromatosis, hereditary hemochromatosis protein, hereditary hemochromatosis protein HLA-H, HH, high Fe, HLAH, HLA-HHFE1, MGC103790, MHC class I-like protein HFE, MVCD7 | |
| A synthesized peptide derived from human HFE (Uniprot #: Q30201) | |
| 50 μg | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cells | |
| 3077 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction