missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HFE Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | HFE |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631506
|
Novus Biologicals
NBP2-99531-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677905
|
Novus Biologicals
NBP2-99531-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HFE Polyclonal antibody specifically detects HFE in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| HFE | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction, Stem Cells | |
| 0.2 um filtered solution in PBS | |
| 3077 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| dJ221C16.10.1, hemochromatosis, hereditary hemochromatosis protein, hereditary hemochromatosis protein HLA-H, HH, high Fe, HLAH, HLA-HHFE1, MGC103790, MHC class I-like protein HFE, MVCD7 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HFE | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title