Learn More
Erlab HF4 U16
Description
- Filter for your air purifier 35 P
- Type of filter: Particulate (ULPA)
- Dimensions: 757,5 x 406,5 x 89 mm
- Quantity/Set: 1
Specifications
Specifications
| For Use With (Equipment) | Fume Hood |
| Product Type | HEPA Filter |