Learn More
Erlab HF4 H14
Product Code. 16822903
Click to view available options
For Use With (Equipment):
Fume Hood
Unit Size:
Each
Product Code. 16822903
Brand: Erlab H11014031
Filter for your air purifier Halo 35 P.
  • Filter for your air purifier 35 P
  • Type of filter: Particulate (HEPA)
  • Dimensions: 757,5 x 406,5 x 89 mm
  • Quantity/Set: 1
Specifications
For Use With (Equipment) Fume Hood
Product Type HEPA Filter