missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEYL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15679-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The sequence of the encoded protein contains a conserved bHLH and orange domain, but its YRPW motif has diverged from other HESR family members. It is thought to be an effector of Notch signaling and a regulator of cell fate decisions. Alternatively spliced transcript variants have been found, but their biological validity has not been determined.Specifications
| HEYL | |
| Polyclonal | |
| Unconjugated | |
| HEYL | |
| bHLHb33, HEY3, HEYL, hHeyL, hHRT3, HRT 3, HRT3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26508, 313575, 56198 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9DBX7, Q9NQ87 | |
| Heyl | |
| HEYL Fusion Protein Ag8163 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction