missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEYL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116766
This item is not returnable.
View return policy
Description
Positive Samples: A-549, BxPC-3 Immunogen sequence: MKRPKEPSGS DGESDGPIDV GQEGQLSQMA RPLSTPSSSQ MQARKKHRGI IEKRRRDRIN SSLSELRRLV PTAFEKQGSS KLEKAEVLQM TVDHLKMLHA.
This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The sequence of the encoded protein contains a conserved bHLH and orange domain, but its YRPW motif has diverged from other HESR family members. It is thought to be an effector of Notch signaling and a regulator of cell fate decisions. Alternatively spliced transcript variants have been found, but their biological validity has not been determined.
Specifications
| HEYL | |
| Polyclonal | |
| Unconjugated | |
| HEYL | |
| bHLHb33; Class B basic helix-loop-helix protein 33; hairy and enhance of split related 3; hairy and enhancer of split-related protein 3; hairy/enhancer-of-split related with YRPW motif 3; hairy/enhancer-of-split related with YRPW motif-like; hairy/enhancer-of-split related with YRPW motif-like protein; Hairy-related transcription factor 3; hes related family bHLH transcription factor with YRPW motif-like; hesr3; hes-related family bHLH transcription factor with YRPW motif-like; HEY3; HEYL; HEY-like protein; hHeyL; hHRT3; Hrt3; HRT-3; mHRT3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 26508, 56198 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.77 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q9DBX7, Q9NQ87 | |
| HEYL | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human HEYL (NP_0553862). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction