missing translation for 'onlineSavingsMsg'
Learn More

HEY2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen HEY2
Concentration 0.3 mg/mL
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16817083
View Documents
Proteintech
10597-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16807083
View Documents
Proteintech
10597-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is a rabbit polyclonal antibody raised against full length of human HEY2.

Downstream effector of Notch signaling which may be required for cardiovascular development. Transcriptional repressor which binds preferentially to the canonical E box sequence 5′-CACGTG-3′. Represses transcription by the cardiac transcriptional activators GATA4 and GATA6.
Specifications

Specifications

HEY2
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Rabbit
Rat, Mouse, Human
Q9QUS4, Q9UBP5
15214, 155430, 23493
HEY2 Fusion Protein Ag0913
Primary
-20°C
HEY2
0.3 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
bHLHb32, CHF1, GRIDLOCK, GRL, hCHF1, HERP, HERP1, HESR 2, HESR2, HEY2, hHRT2, HRT 2, HRT2, Protein gridlock homolog
HEY2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.