missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEY2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567365
This item is not returnable.
View return policy
Description
Immunogen sequence: PCEETTSESDM DETIDVGSEN NYSGQSTSSV IRLNSPTTTS QIMARKK Highest antigen sequence identity to the following orthologs - mouse 88%, rat 90%.
Downstream effector of Notch signaling which may be required for cardiovascular development. Transcriptional repressor which binds preferentially to the canonical E box sequence 5'-CACGTG-3'. Represses transcription by the cardiac transcriptional activators GATA4 and GATA6.
Specifications
| HEY2 | |
| Polyclonal | |
| Unconjugated | |
| Hey2 | |
| basic helix-loop-helix transcription factor; bHLHb32; cardiovascular basic helix-loop-helix factor 1; cardiovascular helix-loop-helix factor 1; Chf1; Class B basic helix-loop-helix protein 32; gridlock; GRL; h hHRT2; hairy and enhancer of split related 2; hairy and enhancer of split-related protein 2; hairy/enhancer-of-split related with YRPW motif 2; hairy/enhancer-of-split related with YRPW motif protein 2; Hairy-E(spl)-related with YRPW motif 2; hairy-related transcription factor 2; hCHF1; HERP; Herp1; Herp2; hes related family bHLH transcription factor with YRPW motif 2; hesr2; HESR-2; hes-related family bHLH transcription factor with YRPW motif 2; HES-related repressor protein 1; HES-related repressor protein 2; hey2; hHRT2; Hrt2; HRT-2; mHRT2; protein gridlock; Protein gridlock homolog; zgc:136746 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23493 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBP5 | |
| Hey2 | |
| Recombinant Human HEY2. Recombinant protein control fragment (Product #RP-108022). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction