missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEY1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-47436-25ul
This item is not returnable.
View return policy
Description
HEY1 Polyclonal specifically detects HEY1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| HEY1 | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| basic helix-loop-helix protein OAF1, BHLHb31, Cardiovascular helix-loop-helix factor 2, CHF-2, CHF2hHRT1, Class B basic helix-loop-helix protein 31, Hairy and enhancer of split-related protein 1, hairy/enhancer-of-split related with YRPW motif 1, hairy/enhancer-of-split related with YRPW motif protein 1, Hairy-related transcription factor 1, HERP2HES-related repressor protein 1, HESR-1, HESR1MGC1274, HES-related repressor protein 2, HRT1, HRT-1OAF1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| HEY1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: HLNNYASQREAASGAHAGLGHIPWGTVFGHHPHIAHPLLLPQNGHGNAGTTASPTEPHHQG | |
| 25 μL | |
| DNA Repair, DNA replication Transcription Translation and Splicing, Neuroscience, Transcription Factors and Regulators | |
| 23462 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction