missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hey L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35774-20ul
This item is not returnable.
View return policy
Description
Hey L Polyclonal antibody specifically detects Hey L in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| Hey L | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| BHLHB33, bHLHb33hHeyL, Class B basic helix-loop-helix protein 33, hairy/enhancer-of-split related with YRPW motif 3, hairy/enhancer-of-split related with YRPW motif-like, hairy/enhancer-of-split related with YRPW motif-like protein, Hairy-related transcription factor 3, HEY3, HEY-like protein, hHRT3, HRT3, HRT-3, MGC12623 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human Hey L (NP_055386.2).,, Sequence:, IRYLGVLEGPSSRADPVRIRLLSHLNSYAAEMEPSPTPTGPLAFPAWPWSFFHSCPGLPALSNQLAILGRVPSPVLPGVSSPAYPIPALRTAPLRRATGIILPARRNVLPSRGASSTRRARPLERPATPVPVAPSSRAARSSHIAPLLQSSSPTPPGPTGSAAYVAVPTPNSSSPGPAGRPAGAMLYHSWVSEITEIGAF | |
| 20 μL | |
| Neuroscience | |
| 26508 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction