missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hey L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-69029
This item is not returnable.
View return policy
Description
Hey L Polyclonal antibody specifically detects Hey L in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Hey L | |
| Polyclonal | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| BHLHB33, bHLHb33hHeyL, Class B basic helix-loop-helix protein 33, hairy/enhancer-of-split related with YRPW motif 3, hairy/enhancer-of-split related with YRPW motif-like, hairy/enhancer-of-split related with YRPW motif-like protein, Hairy-related transcription factor 3, HEY3, HEY-like protein, hHRT3, HRT3, HRT-3, MGC12623 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MKRPKEPSGSDGESDGPIDVGQEGQLSQMARPLSTPSSSQMQARKKHRGII | |
| 100 μg | |
| Neuroscience | |
| 26508 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction