missing translation for 'onlineSavingsMsg'
Learn More

Hexylene glycol, MedChemExpress

Product Code. 16468496
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16468496

Brand: Medchem Express HYB0903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Hexylene glycol is a small molecular weight surfactant, widely used as an industrial coating solvent, does not cause adverse health or environmental effects.

Hexylene glycol is a small molecular weight surfactant, widely used as an industrial coating solvent, does not cause adverse health or environmental effects.

Specifications

Chemical Name or Material Hexylene glycol
Physical Form Liquid
CAS 107-41-5
Color Colorless
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C6H14O2
Synonym 2-Methyl-2,4-pentanediol MPD
Solubility Information H2O : 100 mg/mL (846.24 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (846.24 mM)
SMILES CC(O)(C)CC(O)C
Molecular Weight (g/mol) 118.17
Formula Weight 118.17
Percent Purity 99.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.