missing translation for 'onlineSavingsMsg'
Learn More

Hexyl Octyl Phthalate, TRC

Product Code. 30136821
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136821 500 mg 500mg
30011699 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136821 Supplier Toronto Research Chemicals Supplier No. TRCB185420500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61827-62-1
Molecular Formula C22H34O4
Molecular Weight (g/mol) 362.5
Synonym 1,2-Benzenedicarboxylic Acid, 1-Hexyl 2-Octyl Ester, 1,2-Benzenedicarboxylic Acid Hexyl Octyl Ester
IUPAC Name hexyl octyl phthalate
SMILES O=C(OCCCCCCCC)C1=CC=CC=C1C(OCCCCCC)=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C22H34O4/c1-3-5-7-9-10-14-18-26-22(24)20-16-12-11-15-19(20)21(23)25-17-13-8-6-4-2/h11-12,15-16H,3-10,13-14,17-18H2,1-2H3
Formula Weight 362.25
Percent Purity >95%
Chemical Name or Material Hexyl Octyl Phthalate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.